Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCXR Mouse anti-Human, Clone: OTI9C9, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA500830

Catalog No. 50-167-1976


Add to cart

Description

Description

The protein encoded by this gene acts as a homotetramer to catalyze diacetyl reductase and L-xylulose reductase reactions. The encoded protein may play a role in the uronate cycle of glucose metabolism and in the cellular osmoregulation in the proximal renal tubules. Defects in this gene are a cause of pentosuria. Two transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

DCXR
Monoclonal
0.46 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q7Z4W1
DCR, HCR2, HCRII, KIDCR, P34H, PNTSU, SDR20C1, XR
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51181
Immunohistochemistry (Paraffin), Western Blot
OTI9C9
Unconjugated
DCXR
Liquid
DCXR
Full length human recombit protein of human DCXR produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit