Learn More
DDB1 Mouse anti-Human, Clone: 1322, Abnova™
Mouse monoclonal antibody raised against human DDB1.
Manufacturer:  Abnova Corporation MAB12522
Catalog No. 89-934-037
Add to cart
Description
Description
This gene encodes the large subunit of DNA damage-binding protein which is a heterodimer composed of a large and a small subunit. This protein functions in nucleotide-excision repair. Its defective activity causes the repair defect in the patients with xeroderma pigmentosum complementation group E (XPE). However, it remains for mutation analysis to demonstrate whether the defect in XPE patients is in this gene or the gene encoding the small subunit. In addition, Best vitelliform mascular dystrophy is mapped to the same region as this gene on 11q, but no sequence alternations of this gene are demonstrated in Best disease patients. [provided by RefSeq]
Specifications
Specifications
DDB1
1322
Mouse monoclonal antibody raised against human DDB1.
In PBS.
DDBA/UV-DDB1/XAP1/XPCE/XPE/XPE-BF
Mouse
IgG
50 uL
Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
DDB1
A synthetic peptide corresponding to human DDB1.
Protein G purification
RUO
Primary
1642