Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DDIAS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558430
Description
Immunogen sequence: GFTVIDYFHQ LLQTFNFRKL QCDSQAPNNH LLALDHSNSD LSSIYTSDST SDFFKSCSKD TFSKFWQPSL EFTCIVSQLT DNDDFSASEQ SKAFGTLQQN RKS Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 50%.
DDIAS is a protein coding gene.
Specifications
DDIAS | |
Polyclonal | |
Unconjugated | |
DDIAS | |
C11orf82; C21H11orf82; DDIAS; DNA damage induced apoptosis suppressor; DNA damage-induced apoptosis suppressor; DNA damage-induced apoptosis suppressor protein; Nitric oxide-inducible gene protein; noxin; RGD1559690 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
220042 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IXT1 | |
DDIAS | |
Recombinant protein corresponding to Human DDIAS. Recombinant protein control fragment (Product #RP-103697). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction