Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDIT4 293T Cell Overexpression Lysate (Denatured), Abnova

Cell transient overexpression lysate for DDIT4

Manufacturer:  Abnova Corporation H00054541T01

Catalog No. 89-002-380


Add to cart

Description

Description

  • DNA-damage-inducible transcript 4
  • Transfected cell line: 293T
  • Plasmid: pCMV-DDIT4 full-length
  • Host: Human
  • Theoretical molecular weight: 25.4kDa
  • Storage buffer: 1X sample buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% bromophenol blue)

Western blotting

Specifications

Specifications

Western Blot
293T
NM_019058
6.8
Transient overexpression cell Lysate was tested with Anti-DDIT4 antibody (H00054541-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
DDIT4 293T Cell Transient Overexpression Lysate(Denatured)
25.4
pCMV-DDIT4 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit