Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDK (FLAG) Rabbit anti-Tag, Polyclonal, liquid, OriGene

Rabbit Polyclonal Antibody
Supplier: OriGene TA150078

Description
Synthetic peptide sequence is DYKDDDDK
DDK (FLAG-tag, FLAG octapeptide, FLAG epitope) is a polypeptide protein tag that can be added to a protein using recombit DNA technology, having the sequence motif DYKDDDDK (where D=aspartic acid, Y=tyrosine, and K=lysine). It is an artificial antigen to which specific, high affinity monoclonal antibodies have been developed and hence can be used for protein purification by affinity chromatography and also can be used for locating proteins within living cells. It has been used to separate recombit, overexpressed protein from wild-type protein expressed by the host organism. It can also be used in the isolation of protein complexes with multiple subunits, because its mild purification procedure tends not to disrupt such complexes. It has been used to obtain proteins of sufficient purity and quality to carry out 3D structure determination by x-ray crystallography.Specifications
DDK (FLAG) | |
Polyclonal | |
Whole serum with 0.05% sodium azide | |
Rabbit | |
100 μL | |
Primary | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ANTI DDK, ANTI-DDK, ANTIDDK, FLAG TAG, ANTI FLAG, ANTI-FLAG, ANTI FLAG ANTIBODY, FLAG ANTIBODY, M2 ANTIBODY, FLAG M2, ANTIBODY FOR FLAG, 4C5, ANTI DDK 4C5, DDK 4C5, ANTI-DDK 4C5, DYKDDDDK TAG, D-TAG ANTIBODY, ANTI D-TAG, ANTI D TAG, ANTI-D TAG, ANTI-MDDK, ANTI-MYC DDK, ANTI-MYC/DDK, MYC/DDK ANTIBODY, MYC DDK ANTIBODY, ANTI MYC DDK, ANTI DDK MYC, ANTI-DDK/MYC, ANTI-DDK MYC, FLAG AB, FLAG MAB, DDK ANTIBODY | |
Synthetic peptide conjugated to KLH | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction