Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDO Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA524725

Catalog No. PIPA524725


Only null left
Add to Cart

Description

Description

DDO Polyclonal Antibody for Western Blot, IHC (P)

DDO is a peroxisomal flavoprotein that catalyzes the oxidative deamination of D-aspartate and N-methyl D-aspartate. Flavin adenine dinucleotide or 6-hydroxyflavin adenine dinucleotide can serve as the cofactor in this reaction.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DDO
Polyclonal
Unconjugated
DDO
5330420D20Rik; 5730402C02Rik; AI467244; aspartic oxidase; DASOX; D-aspartate oxidase; D-aspartate oxidase, DDO; Ddo; DDO protein; DDO-1; DDO-2
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
70503, 8528
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.45 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q922Z0, Q99489
DDO
KLH conjugated synthetic peptide between 102-130 amino acids from the central region of human DDO.
400 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.