Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDOST Monoclonal Antibody (OTI2B4)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526611

Catalog No. PIMA526611


Only null left
Add to Cart

Description

Description

DDOST Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a component of the oligosaccharyltransferase complex which catalyzes the transfer of high-mannose oligosaccharides to asparagine residues on nascent polypeptides in the lumen of the rough endoplasmic reticulum. The protein complex co-purifies with ribosomes. The product of this gene is also implicated in the processing of advanced glycation end products (AGEs), which form from non-enzymatic reactions between sugars and proteins or lipids and are associated with aging and hyperglycemia.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DDOST
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P39656
DDOST
Human recombinant protein fragment corresponding to amino acids 131-378 of DDOST produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI2B4
Unconjugated
DDOST
advanced glycation endproduct receptor 1; advanced glycation end-product receptor 1; AGER1; AGE-R1; CDG1R; Ddost; DDOST 48 kDa subunit; dolichyl-diphosphooligosaccharide protein glycosyltransferase; dolichyl-diphosphooligosaccharide-protein glycosyltransferase; dolichyl-diphosphooligosaccharide--protein glycosyltransferase 48 kDa subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit (non-catalytic); dolichyl-di-phosphooligosaccharide-protein glycotransferase; dolichyl-diphosphooligosaccharide-protein glycotransferase; KIAA0115; MGC2191; OK/SW-cl.45; OKSWcl45; oligosaccharyl transferase 48 kDa subunit; oligosaccharyltransferase 48 kDa subunit; oligosaccharyltransferase OST48; oligosaccharyltransferase subunit 48; OST; OST48; WBP1; wu:fa11d01; zgc:66068; zgc:66068 protein; zgc:77478
Mouse
Affinity Chromatography
RUO
1650
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.