Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ DDR1 Recombinant Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$287.00 - $1383.00

Specifications

Formulation PBS (pH 7.4) containing 10% glycerol. containingno Preservative
Gene Alias CD167 antigen-like family member A, CD167a antigen, DDR, discoidin domain receptor family, member 1, discoidin domain receptor tyrosine kinase 1, Discoidin receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.1, EDDR1, ENTRK4, Epithelial discoidin domain recept
Gene ID (Entrez) 780
Gene Symbol DDR1
Name Recombinant DDR1 Protein
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB6022110UG View Documents Novus Biologicals™
NBP26022110UG
10μg Each for $287.00
Add to cart
 
NB6022150UG View Documents Novus Biologicals™
NBP26022150UG
50μg Each for $1,383.00
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal hIgG-His-tag and corresponding to the amino acids 21-417 of Human DDR1 The Recombinant Human DDR1 Protein is derived from Baculovirus. The Recombinant Human DDR1 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

PBS (pH 7.4) containing 10% glycerol. containingno Preservative
780
Recombinant DDR1 Protein
Baculovirus
0.5mg/mL
SDS-Page
CD167 antigen-like family member A, CD167a antigen, DDR, discoidin domain receptor family, member 1, discoidin domain receptor tyrosine kinase 1, Discoidin receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.1, EDDR1, ENTRK4, Epithelial discoidin domain recept
DDR1
Recombinant Protein
RUO
< 1.0 EU per 1ug of protein (determined by LAL method)

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit