Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DDT Monoclonal Antibody (OTI1H3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526176
Description
DDT Monoclonal Antibody for Western Blot, IHC (P)
Tautomerization of D-dopachrome with decarboxylation to give 5,6-dihydroxyindole (DHI).
Specifications
DDT | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P30046 | |
DDT | |
Full length human recombinant protein of DDT produced in HEK293T cell. | |
100 μL | |
Primary | |
Human, Monkey | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1H3 | |
Unconjugated | |
DDT | |
C78655; DDCT; D-dopachrome decarboxylase; D-dopachrome tautomerase; Ddt; dopachrome isomerase; hCG_41098; phenylpyruvate tautomerase II | |
Mouse | |
Affinity Chromatography | |
RUO | |
1652 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction