Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDT Monoclonal Antibody (OTI3A11)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526174

Catalog No. PIMA526174


Only null left
Add to Cart

Description

Description

DDT Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

Tautomerization of D-dopachrome with decarboxylation to give 5,6-dihydroxyindole (DHI).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DDT
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P30046
DDT
Full length human recombinant protein of DDT produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI3A11
Unconjugated
DDT
C78655; DDCT; D-dopachrome decarboxylase; D-dopachrome tautomerase; Ddt; dopachrome isomerase; hCG_41098; phenylpyruvate tautomerase II
Mouse
Affinity chromatography
RUO
1652
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.