Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDT Mouse anti-Human, Monkey, Janelia Fluor™ 549, Clone: OTI1H3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270562JF549

Catalog No. N270562J549


Add to cart

Description

Description

DDT Monoclonal antibody specifically detects DDT in Human, Monkey samples. It is validated for Immunohistochemistry.
Specifications

Specifications

DDT
OTI1H3
Immunohistochemistry
Affinity Purified, Conjugated
DDT
Full length recombinant protein of human DDT (NP_001346) produced in HEK293T cells.
Immunogen affinity purified
Stem Cell Markers
Primary
1652.0
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
D-dopachrome decarboxylase, D-dopachrome tautomeraseDDCT, EC 4.1.1.84, Phenylpyruvate tautomerase II
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Monkey
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit