Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX1 Antibody (3E5E2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP261745
Description
DDX1 Monoclonal specifically detects DDX1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
DDX1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin 1:200-1:1000 | |
ATP-dependent RNA helicase DDX1, DBP-RBDEAD box polypeptide 1, DEAD (Asp-Glu-Ala-Asp) box polypeptide 1, DEAD box protein 1, DEAD box protein retinoblastoma, DEAD box-1, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 1, EC 3.6.1, EC 3.6.1.10, EC 3.6.4.13, UKVH5d | |
Mouse | |
82.4 kDa | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
3E5E2 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
DDX1 | |
Purified recombinant fragment of human DDX1 (AA: 642-740) expressed in E. Coli. | |
Protein G purified | |
Cancer, Neuroscience, Transcription Factors and Regulators, Vision | |
1653 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction