Learn More
DDX10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 178571AP
Catalog No. 50-173-1836
Add to cart
Description
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, and it may be involved in ribosome assembly. Fusion of this gene and the nucleoporin gene, NUP98, by inversion 11 (p15q22) chromosome translocation is found in the patients with de novo or therapy-related myeloid maligcies.
Specifications
Specifications
DDX10
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q13206, Q80Y44
DDX10, DEAD box protein 10, HRH J8
Rabbit
IgG
150 μL
-20°C
Primary
1662, 300710, 77591
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DDX10
Liquid
DDX10
DDX10 Fusion Protein Ag12317
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat