Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX18 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen DDX18
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB126552
View Documents
Novus Biologicals
NBP31082525UL
25 μg
Each for $106.19
Add to cart
 
NB126551
View Documents
Novus Biologicals
NBP310825100UL
100 μg
Each for $352.89
Add to cart
 
Description

Description

DDX18 Polyclonal antibody specifically detects DDX18 in Human samples. It is validated for Western Blot
Specifications

Specifications

DDX18
Western Blot
Purified
RUO
Human
DEAD (Asp-Glu-Ala-Asp) box polypeptide 18, DEAD box protein 18, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 18 (Myc-regulated), EC 3.6.1, EC 3.6.4.13, FLJ33908, MrDbATP-dependent RNA helicase DDX18, Myc-regulated DEAD box protein
The immunogen is a synthetic peptide directed towards the C terminal region of human DDX18 (NP_006764.3). Peptide sequence VALSFGFKVPPFVDLNVNSNEGKQKKRGGGGGFGYQKTKKVEKSKIFKHI
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Core ESC Like Genes, Stem Cell Markers
PBS buffer, 2% sucrose
8886
Polyclonal
Affinity purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit