Learn More
DDX3 Mouse anti-Human, Alexa Fluor™ 750, Clone: 11F11D10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP214848AF750
Catalog No. NP214848A70
Add to cart
Description
Description
DDX3 Monoclonal antibody specifically detects DDX3 in Human samples. It is validated for Western Blot,Flow Cytometry.
Specifications
Specifications
DDX3
11F11D10
Western Blot, Flow Cytometry
DBXCAP-Rf, DDX14, DDX3DEAD/H box-3, DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, X-linked, DEAD box protein 3, X-chromosomal, DEAD box, X isoform, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3, EC 3.6.1, EC 3.6.4.13, helicase like protein 2, Helicase-like protein 2, HLP2ATP-dependent RNA helicase DDX3X
Mouse
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
DDX3X
A partial recombinant protein containing amino acids 24-203 of human DDX3 was used as immunogen for this antibody.
0.1 mL
Primary
1654