Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX3X Mouse anti-Human, Clone:3B9G8, Abnova™

Mouse monoclonal antibody raised against recombinant human DDX3X.

Manufacturer:  Abnova Corporation MAB17182

Catalog No. 89-333-605


Add to cart

Description

Description

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which interacts specifically with hepatitis C virus core protein resulting a change in intracellular location. This gene has a homolog located in the nonrecombining region of the Y chromosome. The protein sequence is 91% identical between this gene and the Y-linked homolog. [provided by RefSeq]
Specifications

Specifications

DDX3X
3B9G8
DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, X-linked
In PBS (0.05% sodium azide)
Liquid
Mouse
IgG1
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Flow Cytometry, Western Blot
Unconjugated
ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
DDX3X
DBX, DDX14, DDX3, HLP2
Recombinant protein corresponding to amino acid 518-661 of human DDX3X from E. coli.
100μg
Antibody
1654
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit