Learn More
Abnova™ DDX3Y 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00008653T01
Catalog No. 89-941-675
Add to cart
Specifications
Specifications
Western Blot
293T
NM_004660.2
pCMV-DDX3Y full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
DDX3Y 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 72.71kDa
Transient overexpression cell lysate was tested with anti-DDX3Y antibody (Mfr. No. H00008653-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)