Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ DDX41 HEK293 Cell Transient Overexpression Lysate (Nondenatured)
Human overexpression lysate
Supplier: Abnova™ L067T6
Specifications
Immunoprecipitation, Western Blot | |
DDX41 HEK293 Cell Transient Overexpression Lysate(Non-Denatured) | |
Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. | |
pCMV-DDX41 full length | |
100 μg | |
Human | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
HEK293 | |
NM_016222 | |
Theoretical M.W. 70kDa | |
Transient overexpression cell lysate was tested with anti-DDX41 antibody (Mfr. No. H00051428-M01) by Western Blot. | |
In modified RIPA Lysis Buffer. | |
2 mg/mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction