Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX42 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549524

Catalog No. PA549524

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a member of the Asp-Glu-Ala-Asp (DEAD) box protein family. Members of this protein family are putative RNA helicases, and are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. Two transcript variants encoding the same protein have been identified for this gene.
Specifications

Specifications

DDX42
Polyclonal
PBS with 0.09% sodium azide
Q86XP3, Q810A7
ATP-dependent RNA helicase DDX42, DEAD box protein 42, RNA helicase-like protein, RHELP, RNA helicase-related protein, RNAHP, SF3b DEAD box protein, Splicing factor 3B-associated 125 kDa protein, SF3b125, DDX42
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11325, 72047
Western Blot
Unconjugated
DDX42
Liquid
DDX42
KLH conjugated synthetic peptide between 104-138 amino acids from human DDX42.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit