Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX56 Mouse anti-Human, Clone: OTI5C1, lyophilized, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene CF802817
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The protein encoded by this gene shows ATPase activity in the presence of polynucleotides and associates with nucleoplasmic 65S preribosomal particles. This gene may be involved in ribosome synthesis, most likely during assembly of the large 60S ribosomal subunit.Specifications
DDX56 | |
Monoclonal | |
Unconjugated | |
DDX56 | |
Lyophilized | |
DDX56 | |
Human recombit protein fragment corresponding to amino acids 323-547 of human DDX56 produced in E.coli. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Immunohistochemistry (Paraffin), Western Blot | |
OTI5C1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NY93 | |
DDX21, DDX26, NOH61 | |
Mouse | |
IgG1 | |
100 μg | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
54606 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title