Learn More
DDX56 Mouse anti-Human, Clone: OTI5A9, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF802823
Catalog No. 50-166-6182
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The protein encoded by this gene shows ATPase activity in the presence of polynucleotides and associates with nucleoplasmic 65S preribosomal particles. This gene may be involved in ribosome synthesis, most likely during assembly of the large 60S ribosomal subunit.
Specifications
Specifications
DDX56
Monoclonal
Unconjugated
DDX56
Lyophilized
DDX56
Human recombit protein fragment corresponding to amino acids 323-547 of human DDX56 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI5A9
PBS with 8% trehalose and no preservative; pH 7.3
Q9NY93
DDX21, DDX26, NOH61
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
54606