Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX6 Mouse anti-Human, Clone: OTI2D6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526662

Catalog No. PIMA526662


Add to cart

Description

Description

This gene encodes a member of the DEAD box protein family. The protein is an RNA helicase found in P-bodies and stress granules, and functions in translation suppression and mRNA degradation. It is required for microRNA-induced gene silencing.
Specifications

Specifications

DDX6
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P26196
HLR2, P54, RCK, ATP-dependent RNA helicase p54, DEAD box protein 6, DEAD box-6, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 6 (RNA helicase, 54kD), oncogene RCK, probable ATP-dependent RNA helicase DDX6
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1656
Immunohistochemistry (Paraffin), Western Blot
OTI2D6
Unconjugated
DDX6
Liquid
DDX6
Human recombit protein fragment corresponding to amino acids 1-298 of DDX6 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit