Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX60L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548242
Description
DDX60L Polyclonal antibody specifically detects DDX60L in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
DDX60L | |
Polyclonal | |
Unconjugated | |
DDX60L | |
DDX60L; DEAD (Asp-Glu-Ala-Asp) box polypeptide 60-like; DEAD box protein 60-like; DEAD-box helicase 60-like; probable ATP-dependent RNA helicase DDX60-like; putative ATP-dependent RNA helicase DDX60 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
91351 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 0.09% sodium azide | |
Q5H9U9 | |
DDX60L | |
KLH conjugated synthetic peptide between 1200-1228 amino acids from the C-terminal region of human DDX60L. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction