Learn More
Deb USA 44 Protective Cream
Manufacturer:  Deb USA 44135
Catalog No. 19-013168
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
6.3 to 7.3
Ideal for manufacturing, chemical and printing facilities, hospitals and healthcare facilities, beauty salons
Off-white to grey
5 oz.
Tube
Cream
44 Protective Cream
Water, Kaolin, Microcrystalline Wax, Paraffinum Liquidum, Paraffin, Stearic Acid, Cetearyl Alcohol, Cocamide DEA, Ceresin, Triethanolamine, Methylparaben, Chloroxylenol, Fragrance, Propylparaben, Acrylic Copolymer
147mL
Each