Learn More
Deb USA AeroGreen Antibacterial Soap
Antibacterial foam hand wash with Triclosan improves skin hydration
Manufacturer:  Deb USA 57250
Catalog No. 19-065-134
Add to cart
Includes: Eight 1L (33 oz.) Proline cartridges
Description
Description
  • High-quality antibacterial formula providing effective protection from most common bacteria
  • Ultrasonically sealed cartridges prevent the ingress of bacteria
  • No water required to create a lather
  • Provides up to 1400 hand washes from each cartridge
  • Contains Triclosan to fight bacteria growth and added moisturizers to keep hands healthy
  • Rinses easily from the skin leaving no soapy residue
  • Only a small amount is required to provide efficient cleaning
Specifications
Specifications
Safety
1L
8 Case
Green
Liquid
5 to 7
0.26 gal.
Cartridge
Triclosan 0.3%, Water, Sodium Laureth Sulfate, Propylene Glycol, PPG-24-Glycereth-24, Cocamidopropyl Betaine, Tetrasodium EDTA, 2-Bromo-2-nitropropane-1,3-diol, Fragrance, Citric Acid, green 5, yellow 5
AeroGreen™ Antibacterial Foam Soap
Ideal for general industry, healthcare, schools, manufacturing and office environments