Learn More
Deb USA FOAM WASH Washroom Cleansers
Pleasantly fragranced, rich and mild foam soap with added moisturizers
Supplier: Deb USA AZU2LT
Description
- Cartridge is ultrasonically sealed, non-vented, and fully recyclable (HDPE Type 2)
- A high quality quick rinsing foam hand wash formula leaves the skin feeling silky smooth with no residue
- Contains skin conditioners to prevent drying of the skin leaving hands feeling soft and supple
- Only one dose, .7 mL, is required to provide effective skin cleansing
- Rose-fragranced
Specifications
Foam Wash Cleanser | |
Liquid | |
Water, Sodium Laureth Sulfate, Propylene Glycol, PEG-75 Lanolin, Cocamidopropyl Betaine, Hydroxypropyl Guar Hydroxypropyltrimonium Chloride, Fragrance, Benzyl Alcohol, Citric Acid, Methylchloroisothiazolinone, Methylisothiazolinone, blue 1 | |
Cartridge | |
4 Pk. | |
2 L |
Blue | |
Green Seal, Federal Food, Drug, and Cosmetic Act | |
For general wash cleanse room Use | |
6 to 7 | |
0.52 gal. |
Safety and Handling
ShelfLife : 2 Years