Learn More
Deb USA FOAM WASH Washroom Cleansers
Pleasantly fragranced, rich and mild foam soap with added moisturizers
Manufacturer:  Deb USA RFW1L
 View more versions of this product
Catalog No. 19-165-466
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Green Seal, Federal Food, Drug, and Cosmetic Act
Water, Sodium Laureth Sulfate, Propylene Glycol, Aloe Barbadensis Leaf Juice, Cocamidopropyl Betaine, PEG-75 Lanolin, Allantoin, Fragrance, Hydroxypropyl Guar Hydroxypropyl trimonium Chloride, Benzyl Alcohol, Citric Acid, Methylchloroisothiazolinone, Methylisothiazolinone, red 40, blue 1
Liquid
Rose FOAM WASH; 1L Cartridge
5 to 6
1L
6 Case
Pink
For general wash cleanse room Use
Rose
Rose Foam Wash Cleanser
0.26 gal.
Cartridge
Safety and Handling
ShelfLife : 2 Year(s)