Deb USA Heavy-Duty Hand Cleansers

Manufacturer: Deb USA ULT150W

 View more versions of this product

Catalog No. 19-165-483

  Check Availability
Add to Cart

    Specifications

    Packaging Pail
    Certifications/Compliance ISO 9001
    Composition Aqua, Alcohol Denatured, Dimethyl Glutarate, Glycerin, Dimethyl Succcinate, Dimethyl Adipate, Trideceth-5, Perfume, Citric Acid, Iodopropynyl Butylcarbamate, Phenoxyethanol, PEG-4, PEG-4 Dilaurate, PEG-4 Laurate, Tetrasodium Iminodisuccinate, Methylparaben, Ethylparaben, Butylparaben, Isobutylparaben, Propylparaben
    Format Wipe
    Shelf Life 24 Month(s)
    Quantity 4 Case
    For Use With (Application) For removing paints, inks, Resins, adhesives and most other print and paint-shop contaminants
    Product Type Ultra Wipes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit