Deb USA InstantFOAM™ Non-Alcohol Free Hand Sanitizer

Instand foam formula sanitizes without drying out skin

Manufacturer: Deb USA 55851

 View more versions of this product

Catalog No. 19-165-455

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Size (English) 0.012 gal.
    Size (Metric) 47mL
    Packaging Bottle, one pump
    Quantity 12 Case
    Certifications/Compliance NSF E-3 rated, Federal Food, Drug, and Cosmetic Act
    Composition Benzalkonium Chloride, Water, Propylene Glycol, Aloe, Barbadensis Leaf Juice, Cocamidopropyl Betaine, Lauramine Oxide, Tetrasodium EDTA, Magnesium Nitrate, Methylchloroisothiazolinone, Magnesium Chloride, Methylisothiazolinone
    For Use With (Application) Ideal for use outside the washroom to provide hand hygiene at the work-point or on the move
    Shelf Life 36 Month(s)
    Product Type Hand Sanitizer

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit