Learn More
Deb USA InstantFOAM™ Non-Alcohol Free Hand Sanitizer
Instand foam formula sanitizes without drying out skin
Manufacturer:  Deb USA 55857
 View more versions of this product
Catalog No. 19-165-456
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
NSF E-3 rated, Federal Food, Drug, and Cosmetic Act
Ideal for use outside the washroom to provide hand hygiene at the work-point or on the move
0.26 gal.
Cartridge
Benzalkonium Chloride, Water, Propylene Glycol, Aloe, Barbadensis Leaf Juice, Cocamidopropyl Betaine, Lauramine Oxide, Tetrasodium EDTA, Magnesium Nitrate, Methylchloroisothiazolinone, Magnesium Chloride, Methylisothiazolinone
Hand Sanitizer
1L
6 Case
Safety and Handling
ShelfLife : 36 Month(s)