Deb USA POWER WASH Heavy Duty Hand Cleansers

Solvent free, natural sunflower oil extracts with cornmeal and ground olive oil stone scrub provides a highly effective deep-clean performance

Manufacturer: Deb USA NPW2LT

 View more versions of this product

Catalog No. 19-165-476

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance EcoLogo and USDA BioPreferred Designated
    Solution Type Natural POWER WASH; 2L Cartridge
    Size (English) 0.52 gal.
    Size (Metric) 2L
    Packaging Cartridge
    Quantity 4 Case
    Color Beige
    Composition Water, Methyl Sunflowerseedate, Olea europaea (Olive) Seed Powder, Trideceth-10, Zea Mays (Corn) Kernel Meal, Glycereth-17 Cocoate, Trideceth-6, Glycerin, Acrylates/C10-30 Alkyl Acrylate Crosspolymer, PEG-200 Hydrogenated Glyceryl Palmate, DMDM Hydantoin, Aminomethyl Propanol, Fragrance, Styrene/Acrylates Copolymer,PEG-7 Glyceryl Cocoate, Tetrasodium Iminodisuccinate, Polyquaternium-7, Benzyl Alcohol, Methylchloroisothiazolinone, Methylisothiazolinone
    For Use With (Application) Ideal for heavy industrial, general and light industrial applications
    Format Liquid
    Shelf Life 24 Month(s)
    Product Type Natural Power Wash Hand Cleanser
    pH Range 6 to 8

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit