Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deb USA Pure RESTORE™ Conditioning Cream

$151.30

Specifications

Color White
Product Type Pure Restore Skin Conditioning Cream
For Use With (Application) For use at the end of each work period to moisturize the skin while natural oils are being replenished, suitable for use on dry, sore, chapped or normal skin
pH Range 6 to7.5
View More Specs

Products 1
Catalog Number Mfr. No. Size (English) Packaging Quantity Size (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Size (English) Packaging Quantity Size (Metric) Price Quantity & Availability  
19-165-465
View Documents Promotion Details
Deb USA
RES1L
33.81 oz. Cartridge 6 Case 999.88mL
Case of 6 for $151.30
Add to cart
 
Description

Description

Specifications

Specifications

White
Pure Restore Skin Conditioning Cream
For use at the end of each work period to moisturize the skin while natural oils are being replenished, suitable for use on dry, sore, chapped or normal skin
6 to7.5
Safety and Handling

Safety and Handling

ShelfLife : 24 Month(s)

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit