Learn More
Deb USA Sanitize InstantFOAM™ Hand Sanitizer
Highly effective perfume and dye free, alcohol-based foam hand sanitizer with moisturizer
Manufacturer:  Deb USA IFS400ML
 View more versions of this product
Catalog No. 19-165-452
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
EcoLogo, BioPreferred Designated and NSF E-3 rated
Ideal for use in any food handling environment
0.1 gal.
Bottle, one pump
Ethyl Alcohol 72%, Water, N-Propanol, Coco-Glucoside, Glyceryl Oleate, PEG-7 Glyceryl Cocoate, PEG-200 Hydrogenated Glyceryl Palmate, Dihydroxypropyl PEG-5 Linoleammonium Chloride, Behentrimonium Chloride, Bis PEG-12, Dimethicone
Instantfoam™ Hand Sanitizer
400mL
6 Case
Safety and Handling
ShelfLife : 36 Month(s)