Deb USA Sanitise InstantFOAM™ Hand Sanitiser

Highly effective perfume and dye free, alcohol-based foam hand sanitizer with moisturizer

Manufacturer: Deb USA IFS50ML

 View more versions of this product

Catalog No. 19-165-451

  Check Availability
Add to Cart

    Specifications

    Size (English) 0.013 gal.
    Size (Metric) 50mL
    Packaging Bottle, one pump
    Quantity 12 Case
    Certifications/Compliance EcoLogo, BioPreferred Designated and NSF E-3 rated
    Composition Ethyl Alcohol 72%, Water, N-Propanol, Coco-Glucoside, Glyceryl Oleate, PEG-7 Glyceryl Cocoate, PEG-200 Hydrogenated Glyceryl Palmate, Dihydroxypropyl PEG-5 Linoleammonium Chloride, Behentrimonium Chloride, Bis PEG-12, Dimethicone
    For Use With (Application) Ideal for use in any food handling environment
    Shelf Life 36 Month(s)
    Product Type Instantfoam™ Hand Sanitizer

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit