Deb USA Sunflower WASH Heavy-Duty Hand Cleanser

Manufacturer: Deb USA SFW4LTR

 View more versions of this product

Catalog No. 19-165-479

  Check Availability
Add to Cart

    Specifications

    Size (Metric) 4L
    Composition Water, Methyl Sunflowerseedate, Trideceth-10, Zea mays (Corn) Kernel Meal, PEG-40 Castor Oil, Laureth-7, Propylene Glycol, Acrylates/C10-30 Alkyl Acrylate Crosspolymer, Fragrance, Diazolidinyl Urea, Methylparaben, Propylparaben, Titanium Dioxide, Sodium Hydroxide, Pentaerythrityl Tetra-di-t-butyl Hydroxyhydro cinnamate, Polyquaternium-7, Magnesium Nitrate, Methylchloroisothiazolinone, Magnesium Chloride, Methyisothiazolinone
    For Use With (Application) Ideal for most common industrial soiling
    Format Liquid
    Shelf Life 24 Month(s)
    Product Type Sunflower Wash Hand Cleanser
    pH Range 6 to 8
    Size (English) 1.05 gal.
    Packaging Cartridge
    Quantity 4 Case
    Certifications/Compliance EcoLogo, Federal Food, Drug, and Cosmetic Act
    Color Cream to light beige

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit