Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deb USA Universal Protect Pre-work Skin Cream

$240.00

Specifications

Color Pale yellow
Product Type Universal Protect Skin Cream
For Use With (Application) Helps protect the skin against contact with various workplace contaminants and working environments such as wet or dry condition
pH Range 6 to7.5
View More Specs

Products 1
Catalog Number Mfr. No. Size (English) Packaging Quantity Size (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Size (English) Packaging Quantity Size (Metric) Price Quantity & Availability  
19-165-481
View Documents Promotion Details
Deb USA
UPW1L
33.81 oz. Cartridge 6 Case 999.88mL
Case of 6 for $240.00
Add to cart
 
Description

Description

Specifications

Specifications

Pale yellow
Universal Protect Skin Cream
Helps protect the skin against contact with various workplace contaminants and working environments such as wet or dry condition
6 to7.5
Safety and Handling

Safety and Handling

ShelfLife : 24 Month(s)

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit