Deb USA Universal Protect Pre-work Skin Cream

Manufacturer: Deb USA UPW150ML

 View all options for this product

Catalog No. 19-165-480


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Size (English) 5 oz.
    Size (Metric) 147mL
    Packaging Tube
    Quantity 12 Case
    Color Pale yellow
    Composition Water, Kaolin, Cetearyl Alcohol, Dicaprylyl Carbonate, Glycerin, Xanthan Gum, Phenoxyethanol, Sodium Cetearyl Sulfate, Sodium Lauryl Sulfate, Methylparaben, 2-Bromo-2-Nitropropane-1,3-diol, Butylparaben, Ethylparaben, Isobutylparaben, Propylparaben, Yellow 5, Yellow 6
    Product Type Universal Protect Skin Cream
    For Use With (Application) Helps protect the skin against contact with various workplace contaminants and working environments such as wet or dry condition
    Form Cream
    pH Range 6 to7.5
    Shelf Life 24 Month(s)

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit