Deb USA Light Duty Hand Cleansers

Manufacturer: Deb USA HAB1L

 View more versions of this product

Catalog No. 19-165-460

  Check Availability
Add to Cart

    Specifications

    Size (English) 0.26 gal.
    Size (Metric) 1L
    Packaging Cartridge
    Quantity 6 Case
    Certifications/Compliance BioPreferred and EcoLogo Certified, Federal Food, Drug, and Cosmetic Act
    Color Pale Green
    Composition Water, Sodium Laureth Sulfate, Sodium Chloride, Cocamidopropyl Betaine, Cocamide MEA, Glycerin, Sodium Cocoamphoacetate, Coco-Glucoside, Fragrance, Citric Acid, DMDM Hydantoin, Polyquaternium-7, Tetrasodium Iminodisuccinate, blue 1, yellow 5
    For Use With (Application) For use in any workplace, leisure or hospitality shower facility
    Format Gel
    Fragrance Rainforest
    Shelf Life 24 Month(s)
    Product Type Hair and Body Cleanser
    pH Range 6 to 8

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit