Deb USA Lime Heavy-Duty Hand Cleanser

Manufacturer: Deb USA LIM4LTR

 View more versions of this product

Catalog No. 19-165-475

  Check Availability
Add to Cart

    Specifications

    Size (English) 1.05 gal.
    Size (Metric) 4L
    Packaging Cartridge
    Certifications/Compliance EcoLogo
    Color Pale green
    Composition Water, Trideceth-10, Corn (Zea mays) Kernel Meal, Glycerin, Trideceth-6, Limonene, Carbomer, Sodium Hydroxide, Triticum Vulgare (Wheat) Germ Oil, 2-Bromo-2-Nitropropane-1,3-Diol, Methylchloroisothiazolinone, Methylisothiazolinone, BHT, yellow 5, blue 1
    For Use With (Application) Removes light greases and oils, ideal for most common industrial soiling
    Format Gritty liquid
    Shelf Life 24 Month(s)
    Product Type Lime Wash Hand Cleanser
    pH Range 7 to 8
    Quantity 4 Case

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit