Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Deb USA UltraGreen Antibacterial Gel
Manufacturer:  Deb USA 75127
 View more versions of this product
Catalog No. 19-999--533
Add to cart
Specifications Description & Specifications
Specifications
Green
Ideal for healthcare, food service, office environments, schools, professional buildings
UltraGreen Antibacterial Gel
0.26 gal.
Cartridge
Triclosan, Water, Sodium Laureth Sulfate, Cocamide DEA, Propylene Glycol, Sodium Chloride, Chloroxylenol, Acrylic Copolymer, Tetrasodium EDTA, Citric Acid, Fragrance, Sodium Hydroxide , Magnesium Nitrate, Methylchloroisothiazolinone, Magnesium Chloride, Methylisothiazolinone, green 5, yellow 5
Gel
6 to 7
1L
8 Case