Deb USA UltraGreen Antibacterial Gel

Manufacturer: Deb USA 75104

 View more versions of this product

Catalog No. 19-999--535

  Check Availability
Add to Cart

    Specifications

    Color Green
    Product Type UltraGreen Antibacterial Gel
    Size (English) 1 gal.
    Size (Metric) 3.78L
    Packaging Cartridge
    Quantity 4 Case
    Composition Triclosan, Water, Sodium Laureth Sulfate, Cocamide DEA, Propylene Glycol, Sodium Chloride, Chloroxylenol, Acrylic Copolymer, Tetrasodium EDTA, Citric Acid, Fragrance, Sodium Hydroxide , Magnesium Nitrate, Methylchloroisothiazolinone, Magnesium Chloride, Methylisothiazolinone, green 5, yellow 5
    For Use With (Application) Ideal for healthcare, food service, office environments, schools, professional buildings
    Format Gel
    pH Range 6 to 7

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit