Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Slice™ Deburring Blade (Concave)

Catalog No. 18222853
Click to view available options
Blade Material:
Zirconium Oxide

Designed for industrial settings, the Slice™ 10484 Deburring Blade (Concave) is an innovative deburring tool that offers increased task efficiency and, depending on your usual tool, reduced risk of injury.

  • Blade type: Concave
  • Lasts up to 11x longer than metal blades
  • Reduces injuries, lowers costs
  • Non-sparking, non-conductive, non-magnetic
  • Chemically inert; never rusts
  • Does not require sharps box disposal
  • 100 percent recyclable
  • Oil and lubricant free
  • 2 single-edge blades per pack

Specifications

Product Type Concave Blade
Blade Material Zirconium Oxide
For Use With (Equipment) Compatible handles: 10482
Dimensions (L x W x H) 60.0 x 15.0 x 1.8 mm
No. per Pack 2 Pk
Weight (Metric) 6.9 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.