Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Decaborane, 98%

In rocket propellants, and as a catalyst in olefin polymerization

Manufacturer:  Alfa Aesar™ 8789206

Quantity 5g

 View more versions of this product

Catalog No. AA8789206


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

17702-41-9
122.22
UN1868
Decaboron tetradecahydride
B10H14
MFCD00011314
14,2845
Decaborane

Specifications

17702-41-9
Powder
213°C
MFCD00011314
UN1868
14,2845
Decaboron tetradecahydride
Slightly soluble in cold water; Hydrolyzes in hot water; Soluble in ethyl acetate, 1-Bromopropane, ethyl silicate, benzene, alcohol, and carbon tetrachloride
98%
0.940g/mL
B10H14
122.22
White to pale yellow
99° to 103°C
5g
Pungent
Decaborane

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228-H301-H310-H330
Flammable solid.
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement
P210-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: DECABORANE

RTECSNumber : HD1400000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit