Alfa Aesar™ Decaborane, 98%

In rocket propellants, and as a catalyst in olefin polymerization

Manufacturer: Alfa Aesar™ 8789214

Quantity 25g

 View more versions of this product

Catalog No. AA8789214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17702-41-9
    Molecular Formula B10H14
    Formula Weight 122.22
    MDL Number MFCD00011314
    UN Number UN1868
    Merck Index 14,2845
    Synonym Decaboron tetradecahydride
    Chemical Name or Material Decaborane

    Specifications

    Formula Weight 122.22
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Odor Pungent
    MDL Number MFCD00011314
    Health Hazard 1 Danger
    Density 0.940g/mL
    RTECS Number HD1400000
    Boiling Point 213°C
    Color White to pale yellow
    UN Number UN1868
    Synonym Decaboron tetradecahydride
    Health Hazard 2 GHS H Statement
    H228-H301-H310-H330
    Flammable solid.
    Toxic if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: DECABORANE
    Physical Form Powder
    CAS 17702-41-9
    Melting Point 99° to 103°C
    Merck Index 14,2845
    Quantity 25g
    Solubility Information Slightly soluble in cold water; Hydrolyzes in hot water; Soluble in ethyl acetate, 1-Bromopropane, ethyl silicate, benzene, alcohol, and carbon tetrachloride
    Molecular Formula B10H14
    Chemical Name or Material Decaborane
    Assay 98%

    GHS H Statement
    H228-H301-H310-H330
    Flammable solid.
    Toxic if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.

    GHS P Statement
    P210-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit