Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Decacarbonyldimanganese, C 30.6%

Manufacturer:  Alfa Aesar™ 1305814

Quantity 25g

 View more versions of this product

Catalog No. AA1305814


  Check Availability
Add to cart

Specifications

Description

  • Starting material for manganese carbonyl chemistry, as a catalyst and an antiknock additive
Description & Specifications

Chemical Identifiers

10170-69-1
389.98
UN3466
Manganese carbonyl; Dimanganese decacarbonyl
Mn2(CO)10
MFCD00011115
14,5727
Decacarbonyldimanganese

Specifications

10170-69-1
Crystalline
80°C (1mmHg) (subl.)
MFCD00011115
UN3466
C 30.6%
25g
Soluble in organic solvents; Insoluble in water
Decacarbonyldimanganese
1.75g/mL
Mn2(CO)10
389.98
Yellow
152° to 154°C
14,5727
Manganese carbonyl; Dimanganese decacarbonyl
Air Sensitive

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement
P280f-P304+P340-P309-P310
Wear protective clothing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit