Alfa Aesar™ Decacarbonyldimanganese, C 30.6%

Manufacturer: Alfa Aesar™ 1305803

Quantity 1g

 View more versions of this product

Catalog No. AA1305803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10170-69-1
    Linear Formula Mn2(CO)10
    Formula Weight 389.98
    MDL Number MFCD00011115
    UN Number UN3466
    Merck Index 14,5727
    Synonym Manganese carbonyl; Dimanganese decacarbonyl
    Chemical Name or Material Decacarbonyldimanganese

    Specifications

    Formula Weight 389.98
    Health Hazard 3 GHS P Statement
    P280f-P304+P340-P309-P310
    Wear protective clothing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Health Hazard 2 GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.
    Boiling Point 80°C (1mmHg) (subl.)
    MDL Number MFCD00011115
    UN Number UN3466
    Melting Point 152° to 154°C
    Sensitivity Air Sensitive
    Density 1.75g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
    TSCA TSCA
    Physical Form Crystalline
    CAS 10170-69-1
    Synonym Manganese carbonyl; Dimanganese decacarbonyl
    Health Hazard 1 Danger
    Color Yellow
    Concentration C 30.6%
    Quantity 1g
    Solubility Information Soluble in organic solvents; Insoluble in water
    Linear Formula Mn2(CO)10
    Merck Index 14,5727
    Chemical Name or Material Decacarbonyldimanganese

    Description

    • Starting material for manganese carbonyl chemistry, as a catalyst and an antiknock additive

    GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.

    GHS P Statement
    P280f-P304+P340-P309-P310
    Wear protective clothing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit