Alfa Aesar™ Decacarbonyldirhenium, 96%

Manufacturer: Alfa Aesar™ 62108MD

Quantity 250mg

 View more versions of this product

Catalog No. AA62108MD

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 14285-68-8
    Molecular Formula C10O10Re2
    Linear Formula Re2(CO)10
    Formula Weight 652.52
    MDL Number MFCD00011198
    UN Number UN3466
    Synonym Dirhenium decacarbonyl; Rhenium carbonyl
    Chemical Name or Material Decacarbonyldirhenium

    Specifications

    Formula Weight 652.52
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 14285-68-8
    UN Number UN3466
    Health Hazard 2 GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.
    Quantity 250mg
    Synonym Dirhenium decacarbonyl; Rhenium carbonyl
    Health Hazard 1 Danger
    TSCA TSCA
    Linear Formula Re2(CO)10
    MDL Number MFCD00011198
    Color White
    Density 2.87g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
    Physical Form Powder
    Molecular Formula C10O10Re2
    Melting Point 170°C (dec.)
    Chemical Name or Material Decacarbonyldirhenium
    Assay 96%

    GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.

    GHS P Statement
    P261-P301+P310-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit