Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decafluorobenzophenone, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
853-39-4
C13F10O
362.13
WWQLXRAKBJVNCC-UHFFFAOYSA-N
decafluorobenzophenone,benzophenone, decafluoro,perfluorobenzophenone,methanone, bis pentafluorophenyl,bis perfluorophenyl methanone,bis pentafluorophenyl methanone,bis 2,3,4,5,6-pentafluorophenyl methanone,di2,3,4,5,6-pentafluorophenyl ketone,acmc-209q5n
70068
bis(2,3,4,5,6-pentafluorophenyl)methanone
C1(=C(C(=C(C(=C1F)F)F)F)F)C(=O)C2=C(C(=C(C(=C2F)F)F)F)F
853-39-4
C13F10O
362.13
WWQLXRAKBJVNCC-UHFFFAOYSA-N
decafluorobenzophenone,benzophenone, decafluoro,perfluorobenzophenone,methanone, bis pentafluorophenyl,bis perfluorophenyl methanone,bis pentafluorophenyl methanone,bis 2,3,4,5,6-pentafluorophenyl methanone,di2,3,4,5,6-pentafluorophenyl ketone,acmc-209q5n
70068
bis(2,3,4,5,6-pentafluorophenyl)methanone
C1(=C(C(=C(C(=C1F)F)F)F)F)C(=O)C2=C(C(=C(C(=C2F)F)F)F)F
Quantity 1g
Packaging Glass bottle
Melting Point 91.0°C to 95.0°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula (C6F5)2CO
Formula Weight 362.13
Percent Purity 97%
Chemical Name or Material Decafluorobenzophenone

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell.

Warning