Alfa Aesar™ Decahydronaphthalene-d{18}, cis + trans, 99% (Isotopic)

Manufacturer: Alfa Aesar™ 4227803

Quantity 1g

 View more versions of this product

Catalog No. AA4227803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 28788-42-3
    MDL Number MFCD00044371
    UN Number UN1147
    Chemical Name or Material Decahydronaphthalene-d{18}, cis + trans, Isotopic

    Specifications

    Physical Form Liquid In Prescored Ampoules
    Health Hazard 3 GHS P Statement
    P260-P280h-P273-P305+P351+P338-P309-P310
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    CAS 28788-42-3
    Density 1.014g/mL
    Boiling Point 70° to 71°C (17mmHg)
    Flash Point 57° (135°F)
    Refractive Index 1.4745
    MDL Number MFCD00044371
    UN Number UN1147
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H226-H315-H319-H335
    Toxic if inhaled.
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DECAHYDRONAPHTHALENE
    Quantity 1g
    Chemical Name or Material Decahydronaphthalene-d{18}, cis + trans, Isotopic
    Assay 99%

    GHS H Statement
    H331-H226-H315-H319-H335
    Toxic if inhaled.
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P260-P280h-P273-P305+P351+P338-P309-P310
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit