Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decanenitrile, 99%, Acros Organics

Click to view available options
Quantity:
500mg
1975-78-6
C10H19N
153.269
MFCD00039505
HBZDPWBWBJMYRY-UHFFFAOYSA-N
decanonitrile,caprinitrile,1-cyanononane,n-decanenitrile,nonyl cyanide,n-nonyl cyanide,unii-1602vq5ivc,1-decanenitrile,nitrile 10 d,decanoic acid nitrile
74791
decanenitrile
CCCCCCCCCC#N
1975-78-6
C10H19N
153.269
MFCD00039505
HBZDPWBWBJMYRY-UHFFFAOYSA-N
decanonitrile,caprinitrile,1-cyanononane,n-decanenitrile,nonyl cyanide,n-nonyl cyanide,unii-1602vq5ivc,1-decanenitrile,nitrile 10 d,decanoic acid nitrile
74791
decanenitrile
CCCCCCCCCC#N
Quantity 500mg
Infrared Spectrum Authentic
Refractive Index 1.4280 to 1.4300 (20°C, 589nm)
Color Colorless
Assay Percent Range 99%
Linear Formula CH3(CH)8CN
Formula Weight 153.27
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Decanenitrile

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning